Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Biology Kit: Simulated Urinalysis
SCI_ED

Catalog No. S06765 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06765 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06765 Supplier Aqua Phoenix Scientific Supplier No. 8306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor Manual
  • Student Data Sheet
  • SDS
  • Normal Urine Sample
  • High Glucose Urine Sample
  • High Albumin Urine Sample
  • High Lead Urine Sample
  • Urine Coloring Solution
  • Sodium Sulfide
  • Unversal Indicator Solution
  • Acetic Acid
  • Glucose Test Strips
  • Microscope Slides
  • Coverslips
  • Empty Dropper Bottles
  • Micropipettes
  • Urine Sample Cups with Lids

Requires

Bunsen burner, balance, microscope, lab glassware, urine hydrometers (optional) and a centrifuge (1500rpm, optional)

Students determine general characteristics of urine sample.

Urinalysis is a valuable procedure used in the health care profession for evaluating human and animal health.

  • Introduces students to biomedical urinalysis procedures similar to those utilized in medical laboratories
  • Includes tests for glucose, albumin, pH, specific gravity and lead
  • Grades 9 to 12
  • Class size: 24 students

No urine or human products are used in this kit.

Specifications

Grade 9 to 12
For Use With (Application) To determine general characteristics of the urine sample
Includes Microscope slides, empty Plastic dropper bottles, pipets, universal color chart, urine sample cups with lids, normal urine sample powder, high Glucose Urine sample powder, high Albumin Urine sample powder, high lead Urine sample powder, coloring solution, Sodium Sulfide solution, universal indicator solution, Acetic Acid solution
Product Type Simulated Urinalysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.