Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Blood Typing with Saliva Kit
SCI_ED

Catalog No. s25080 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S25080 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S25080 Supplier Aqua Phoenix Scientific Supplier No. 3214
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Return cards for type A, B, O red blood cells and H Lectin
  • Anti-A Serum
  • Anti-B Serum
  • MSDS
  • Instructor's manual
  • Student data sheet

Requires

Centrifuge or prolonged settling

Students use their own saliva, from which antigen secretors will be able to determine their ABO blood type.

 

  • Provides real antisera for demonstrating true ABO blood typing agglutination reactions
  • Enhanced instructor's manual with background information, reproducible student protocols and worksheets and complete answer keys
  • Ideal for students in advanced biology, physiology, genetics, forensics, or biochemistry
  • Antisera and blood cells are perishable, refrigeration prolongs shelf life
  • Materials sufficient for up to 50 tests
  • Grades 9-14
  • Class size: 24 students

Materials must ship 2nd day air on ice in summer months. Reply card items are only shipped within the continental United States. Cells should be used within 20 days.

Specifications

Grade 8 to 12
Includes Return cards for vials of a, b and o red blood cells, vials of anti-a and anti-b sera, lectin h, instructor's manual, 30 student handbooks
Product Type Blood Typing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.