Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Chemistry Kit: Characteristics of Matter
SCI_ED

Catalog No. s06736 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
:
Characteristics of Matter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S06736 Characteristics of Matter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06736 Supplier Aqua Phoenix Scientific Supplier No. 1114
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Spot plates, wood stirrers, thermometers, 0.5 oz. plastic bottles with droppers and caps, blue and red litmus, pipets, ammonia solution, baking soda solution, calcium chloride solution, vinegar, washing soda solution, calcium chloride solution, calcium carbonate, sodium chloride, sucrose, sand, instructor manual, special needs guide, methods of assessment manual, SDS

Requires

Balance, glassware and ice.

Explore various physical and chemical properties of matter.

  • Students can observe chemical properties of five solutions by observing their reactions with other solutions
  • Then determine names of two unknown solutions
  • Ten experiments to explore various characteristics of matter: color, odor, solubility, shape, size, texture, density, alkalinity, acidity, states of matter, boiling point and melting point (graphing of melting and boiling points)
  • Grades 5-9
  • Class size: 24 students

Specifications

Grade 1 to 6
For Use With (Application) To explore the characteristics of matter such as color, odor, solubility, density, alkalinity, acidity, states of matter, texture, boiling point, melting point
Includes Spot plates, Wood stirrers, thermometers, 0.5 oz. Plastic bottles with droppers and caps, blue and red litmus, pipets, Ammonia solution, Baking Soda solution, Calcium Chloride solution, Vinegar, Washing Soda solution, Calcium Carbonate, Sodium Chloride, Sucrose, sand, instructor manual, special needs guide, methods of assessment manual, SDS
Product Type Characteristics of Matter Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.