Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Chemistry Kit: Chemical Formulas I
SCI_ED

Catalog No. S06745 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06745 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06745 Supplier Aqua Phoenix Scientific Supplier No. 13122
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Plastic bottles with dropper caps, calcium chloride solution, sodium carbonate solution, copper sulfate solution, sodium hydroxide solution, instructor manual, methods of assessment manual, special needs guide, SDS

Requires

Goggles, test tubes, and test tube racks

Experimentally determine the correct chemical formulas for various compounds.

  • Students combine chemicals in various ratios and observe the combination that produces greatest amount of precipitate
  • Use the ratio of the combination to determine chemical formulas of newly formed substances
  • Includes all required chemicals, vocabulary, chemical formula writing and naming worksheets, data sheets, reproducible procedures, and answer keys
  • Grades 7-12
  • Class size: 24 students

Specifications

Grade 7 to 12
For Use With (Application) For students experimentally determine the correct chemical formulas for various compounds by combining chemicals in appropriate ratios
Includes Plastic bottles with dropper caps, calcium chloride solution, sodium carbonate solution, copper sulfate solution, sodium hydroxide solution, instructor manual, methods of assessment manual, special needs guide, sds
Product Type Chemical Formulas I Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.