Learn More
Description
Includes
Copper Sulfate solution, Hydrochloric Acid solution, Sodium Hydroxide, Acetic Acid, Ammonium Hydroxide, Sodium Carbonate, Sodium Bicarbonate, Unknown A, Unknown B, Unknown C, Unknown D, instructor manual, special needs guide, methods of assessment manual, SDS
Requires
Goggles
- Students identify chemical properties by observing reaction of seven test solutions with other solutions
- Evidence of chemical reactions include gas production, color changes and formation of precipitates
- Students use what they learn about chemical properties to identify four unknown solutions
- Includes all required chemicals, reaction plates, reproducible worksheets and student procedures
- Grades 9-12
- Class size: 24 students
Specifications
Specifications
| Grade | 7 to 12 |
| For Use With (Application) | For students to test chemical properties of 7 different solutions in every possible combination and identify chemical properties by observing their reaction with other solution |
| Includes | Copper Sulfate solution, Hydrochloric Acid solution, Sodium Hydroxide, Acetic Acid, Ammonium Hydroxide, Sodium Carbonate, Sodium Bicarbonate, Unknown A, Unknown B, Unknown C, Unknown D, instructor manual, special needs guide, methods of assessment manual, SDS |
| Product Type | Chemical Properties Chemistry Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
