Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™: Crime Scene
SCI_ED

Catalog No. S06742 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06742 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06742 Supplier Aqua Phoenix Scientific Supplier No. 10542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Student investigation packets, graph paper, glass slides, cover slips, mounting media, universal indicator solution, sifting screen, soil samples, hair, rope samples, molding clay, trash evidence including nine trace evidence samples, plastic spoons, magnifying glasses, instructor manual, SDS

Requires

Microscopes.

Students gather physical evidence to make factual interpretations of the scene.

When suspects and victims enter and/or leave the scene, they bring with them and carry away useful pieces of evidence. Outdoors areas are searched first because changing weather can destroy evidence. Everything must be documented. Court cases have been made by proper documentation and the interpretation of the facts of the processed scene.

  • Students work through six exercises using reasoning, common sense, math skills, biology and chemistry to document crime scene
  • Grades 5-8
  • Class size: 24 students (six groups of four)
Includes:
  • Scene Mapping
  • Hair Analysis
  • Soil Analysis
  • Rope Analysis
  • Analysis of Scene Trash
  • Police Evidence

Specifications

Certifications/Compliance National standards
Grade 5 to 8
For Use With (Application) Scene mapping, hair analysis, soil analysis, rope analysis, analysis of crime scene trash and interpretation of police evidence
Includes Instructor manual, student investigation packets, MSDS, graph Paper, Glass slides, cover slips, mounting media, universal indicator solution, sifting screen, soil samples, hair samples, rope samples, molding clay, trash evidence (including 9 trace evidence samples), Plastic spoons, magnifying Glasses
Requires Microscope, graduated cylinders, test tubes, rulers, markers, notebook paper, white paper and water
Class Size 24 Students
Product Type Crime Scene Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.