Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Dental Cavity Potential Kit
SCI_ED

Catalog No. S25001 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S25001 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S25001 Supplier Aqua Phoenix Scientific Supplier No. 8300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Instructor Manual with Reproducible Student Data Sheet, Plastic Cups, Microcentrifuge Tubes, 5mL, Sodium Hydroxide, Glucose, Glucose Test Strips, Oxidizing Dye, Color Charts

Requires

24 caramels

Kit promotes dental health.

Designed by Temple University's School of Dentistry, this kit allows students to quantitatively determine the bacterial content of their own mouths and to measure the rinsing capacity of their mouths.

  • Provides students with enough information to radically change eating habits in order to prevent tooth decay
  • Students see genuine improvement in their dental health
  • Grades 6-12
  • Class size: 24 students

Specifications

Grade 6 to 12
For Use With (Application) Provides students with enough information to radically change eating habits in order to prevent tooth decay
Includes Oxidizing dye, naoh, glucose, test tape, color charts, 24 three dram vials with caps and one pack of mixing sticks, also includes 24 student data sheets and an instruction manual
Product Type Dental Cavity Potential Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.