Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Dichotomous Keys Kit
SCI_ED

Catalog No. S06767 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Dichotomous Keys Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S06767 Dichotomous Keys Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06767 Supplier Aqua Phoenix Scientific Supplier No. 1178
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Instructor Manual, Student Information Sheets Special Needs Practice with Shapes Watch Pictures Coin Keys Coin Pictures Vehicle Keys Shapes Keys Simplified Tree keys Tree Key Definitions Alternative Sample Keys Evergreen Conifers Key and Master Tree Finder Book

Requires

Colored pencils or crayons.

Guide students through the process of classifying and identifying objects with real-world examples, just like scientists!

  • Using dichotomous keys for beginners or advanced learners
  • Includes seven activities, ranging from easy to challenging, basic to scientific
  • Culminating activities include: Constructing a Tree Key to Your Own School and Using a Dichotomous Key to Identify Seashells
  • Includes real shells for classifying
  • Identify real trees using their scientific names
  • Grades 5-8
  • Class size: 24 students

Specifications

For Use With (Application) Enables students to understand the process of classification and problem solving
Includes Instructor manual, student information sheets special needs practice with shapes watch pictures coin keys coin pictures vehicle keys shapes keys simplified tree keys tree key definitions alternative sample keys evergreen conifers key and master tree finder book
Product Type Dichotomous Keys Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.