Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Elements, Compounds and Mixtures I Kit
SCI_ED

Catalog No. s06453 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06453 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06453 Supplier Aqua Phoenix Scientific Supplier No. 13106
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Filter paper, chromatography paper, black ink, copper sulfate, sodium chloride, copper carbonate, sulfur powder, instructor manual, SDS.

Requires

Goggles, balance.

Introduce students to various concepts of elements, compounds, and mixtures.

  • Special emphasis on separating mixtures by filtration and differences in solubility, chromatography and evaporation
  • Separate two mixtures by filtration and differences in solubility
  • Separate one mixture using paper chromatography
  • Separate two mixtures by evaporation
  • Isolate component colors from a chromatogram to examine individual components
  • Includes all required chemicals, vocabulary exercise, reproducible worksheets and procedures
  • Grades 7-10
  • Class size: 24 students

Specifications

Grade 7 to 12
For Use With (Application) To provide students with a good introduction to chemistry
Includes Instructor manual, special needs guide, sds, terms sheets, an activity sheet, laboratory exercises, a crossword puzzle and a test
Product Type Elements, Compounds and Mixtures I Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.