Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensic Impressions Kit
SCI_ED

Catalog No. S06741 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06741 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06741 Supplier Aqua Phoenix Scientific Supplier No. 10538
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Vials of carbon powder, small paint brushes, plain paper stock, modeling clay, plaster of Paris, teaspoons, five different ropes, magnifying glass, tongue depressors, tire pictures, small ink roller, ink, paper cups, soil, sifting screen, toothbrushes, pans, Petri dishes, instructor's manual, students manual, assessment manual, MSDS

Requires

An old tire, an old gym shoe, mixing containers, a spray bottle and newspaper

Let students become CSI detectives to identify, preserve and evaluate trace evidences at a crime scene.

  • Based on the logic of the exchange principle by Edmund Locard which says, “it is simply impossible to enter a location without changing it in some way, either by bringing something to it or by taking something from it”
  • Students learn to cast six types of impressions using carbon powder, plaster, clay and ink
  • Students understand that footprints or shoe prints, tire tracks, or other impressions imbedded in soft material serve as valuable class evidence for the crime scene investigator
  • Includes six different labs, allowing students to identify and evaluate trace evidence and solve the scenario
  • Grades 8 to 12
  • Class size: 24 students

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) For students become csi detectives and identify and evaluate trace evidence at the crime scene and solve the scenario
Includes Vials of carbon powder, small paint brushes, plain Paper stock, modeling clay, Plaster of Paris, teaspoons, five different ropes, magnifying Glass, tongue depressors, tire impressions pictures, small ink roller, ink, Paper cups, soil, sifting screen, tooth brushes, pans, Petri dishes, instructor's manual, students manual, assessment manual, MSDS
Requires Old tire, an old gym shoe, mixing containers, a spray bottle, and newspaper
Class Size 24 Students
Product Type Forensic Impressions Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.