Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensics Kit: Analysis of Drugs and Poisons
SCI_ED

Catalog No. S25702 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Certifications/Compliance:
National standards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
S25702 National standards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S25702 Supplier Aqua Phoenix Scientific Supplier No. 10502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Instructor, student and Methods of Assessment manuals; student data sheets; special needs materials; Material Safety Data Sheets; universal indicator chart; Q™-Tips; depression plates; Alka Seltzer™; sodium bicarbonate; aspirin; Tylenol™; pipets; plastic bottles with dropper caps; red toothpicks; capillary tubes; chromatography paper; unknowns A and B; unknown powders X, Y and Z; simulated LSD; simulated marijuana; crime scene powder; universal indicator; simulated urine alanine, glycine and unknown; acetic acid; acidified ferric nitrate solution; potassium chromate solution; hydrochloric acid solution; lead nitrate solution; unknown heavy metal; mercury I nitrate solution; butanol; acetone; ninhydrin

Requires

Goggles, UV light, microscopes, hot plate or heat source, balance, glassware and lab supplies

Learn to identify some common over-the-counter drugs

This kit enables students to identify several common over-the-counter drugs through chemical analysis of simulated body fluids. In addition, it teaches students test procedures for controlled substances and heavy metal poisoning as well as amino acids in body fluids as part of a simulated poisoning.

  • Conduct test procedures for “controlled substances”
  • Develop test methods for heavy metal poisoning
  • For qualitative analysis in intriguing context of criminal investigation
  • Grades: 9-14
  • Class size: 24 students

Order Info

Class size: 24 students working in groups.

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) Qualitative analysis in the intriguing context of criminal investigation
Includes Instructor manual with reproducible student data sheet, msds, simulated urine alanine, simulated urine glycine, simulated urine unknown, crime scene unknown, unknown “a”, unknown “b”, unknown powder “x”, unknown heavy metal, unknown powder “Y”, unknown po
Requires UV light, microscope, oven, lamp, or hot plate, scale, glassware and lab supplies
Class Size 24 Students
Product Type Analysis of Drugs and Poisons Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.