Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensics Kit: Analysis of Minerals and Soils
SCI_ED

Catalog No. s25706 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Certifications/Compliance:
National standards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
S25706 National standards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard - Small parts not for children under three years. Contains lead.
Catalog No. S25706 Supplier Aqua Phoenix Scientific Supplier No. 10500
Only null left
Add to Cart
Add to Cart
Choking hazard - Small parts not for children under three years. Contains lead.

Includes

Soil sample A, soil sample B, soil sample C, soil sample D, soil sample Crime Scene, suspect A (glass beads), suspect B (glass beads), crime scene (glass beads), density gradient tubes, medicine droppers, microscope slides, polarizing film, magnifying glasses, plastic forceps, plastic spoons, aluminum pans, sets of sieves, universal indicator chart, gummed labels, bromoform, bromobenzene, universal indicator, potassium chromate solution, hydrochloric acid solution, instructor manual, student data sheets, special needs materials, methods of assessment, manual, MSDS

Requires

Fume hood, UV light, balance, microscope and hot plate.

Student detectives analyze minerals and soils.

Could the suspect have been at the crime scene? To answer this question, students analyze soil, glass, dust, and metal traces using chemical and physical techniques. In addition to providing a fascinating look at one aspect of forensic science, this kit provides students with multiple opportunities to implement the scientific method and better understand the practical uses of science.

  • Designed to teach soil analysis using physical and chemical techniques; comparisons of acidity, particle size, density, and mineral analysis by use of wet chemistry
  • Grades: 9-14
  • Class size: 24 students (six groups of four)

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) For student detectives analyze minerals and soils
Includes Soil sample A, B, C, D, soil sample crime scene, suspect A, suspect B, crime scene, density gradient tubes, medicine droppers, microscope slides, polarizing film, magnifying Glasses, Plastic forceps, Plastic spoons, aluminum pans, sets of sieves, universal indicator chart, gummed labels, Bromoform, Bromobenzene, universal indicator, potassium chromate solution, hydrochloric acid solution, instructor manual, student data sheets, special needs materials, methods of assessment, manual, MSDS
Requires Fume hood, UV light, balance, microscope, oven/lamp/or hot plate, glassware, and general lab supplies
Class Size 24 Students
Product Type Analysis of Mineral and Soil Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.