Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensics Kit: Fabrics and Fiber
SCI_ED

Catalog No. s06739 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06739 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06739 Supplier Aqua Phoenix Scientific Supplier No. 10534
Only null left
Add to Cart
Add to Cart

Includes

Fabric and fiber samples (white cotton, white wool, white carpet fiber, spun glass strands, nylon, rayon, acetate, unknown 1, unknown 2, unknown carpet fiber), wax pencils, metal forceps, plastic forceps, slides, cover slips, wooden toothpicks, 0.5 oz. empty dropping bottles, red litmus paper, lead acetate paper, 3M HCl, 3M NaOH, chlorine bleach, dry-cleaning solvent, acetone, mounting solution, instructor's manual, student manual, MSDS, methods of assessment manual

Requires

Microscopes, alcohol or Bunsen burners.

Examine various types of fabric and fiber evidence and observe the various types of characteristics that are exhibited.

Forensic scientists examine the following:

  • A mineral type (glass wool)
  • Animal type (wool)
  • Vegetable type (cotton)
  • Synthetic types (rayon, nylon and acetate)
  • Observe and test a carpet sample to see the uniqueness of its general overall characteristics and structures
Includes five units:
Experiment No. 1: Animal, Vegetable, Mineral or Fake (The Effect of Heat and Chemical Reaction)
Experiment No. 2: General Microscopic Observation
Experiment No. 3: The Fiber's Reaction to Chemicals (Solubility Test)
Experiment No. 4: What Was the Fiber Used For (Stretch and Strain)
Experiment No. 5: Unknown Fibers and Fabrics

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) For examine various types of fabric and fiber evidence and observe the various types of characteristics that are exhibited
Includes Fabric and Fiber samples white Cotton, white wool, white carpet Fiber, spun Glass, Nylon, Rayon, Acetate, unknown 1, unknown 2, unknown carpet Fiber, Wax pencils, Metal forceps, Plastic forceps, slides, cover slips, wooden toothpicks, 0.5 oz. empty dropping bottles, red litmus Paper, lead Acetate Paper, 3M HCl, 3M NaOH, chlorine bleach, dry-cleaning solvent, Acetone, mounting solution, instructor's manual, student manual, MSDS, methods of assessment manual
Requires Microscope, alcohol or bunsen burners, matches, test tubes, 1-hole stopper, test tube clamp, and distilled water
Class Size 24 Students
Product Type Forensic Chemistry: Fabric and Fiber Analysis Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.