Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensics Kit: Introduction to Toxicology
SCI_ED

Catalog No. s25717 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Certifications/Compliance:
National standards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
S25717 National standards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25717 Supplier Aqua Phoenix Scientific Supplier No. 10530
Only null left
Add to Cart
Add to Cart

Includes

Note on phenylpropanolamine; Rf value cards; tox elution columns; plastic bottles with dropper caps; silica plates; capillary tubes; pipet; acetone; methanol; simulated urine; eluent; ethyl acetate; ammonium hydroxide; potassium iodide solution; acidified methanol; chloroplantinic solution; ninhydrin; diphenyl carbozone; mercuric oxide; Contact™; Sudafed™; Ceffedrine™; Dexatrim™; quinine; sulfuric acid solution, instructor, student and methods of assessment manuals; MSDS

Requires

Goggles, fume hood, both short and long wave UV light, water bath, hair dryer/heat source and ring stands

Demonstrate techniques used to screen for drug use.

  • Emphasizes scientific method principles by careful observation and data collection
  • Includes several forensic scenarios for realistic sleuthing
  • Makes a great applied biology or chemistry lab
  • Provides an opportunity to go beyond the science and discuss the social issues of drug abuse
  • Grades 10-14
  • Class size: 24 students (six groups of four)

Specifications

Certifications/Compliance National standards
Grade 10 to 14
For Use With (Application) For completing the thin layer chromatography and visualization process
Includes Instructor manual, student data sheets, msds, “fbi” fingerprint classification sheets, bond paper, filter paper, gloves, pipet, fingerprint brushes, magnifying glasses, forceps, jars with caps, metal spoons, aluminum foil, superglue, ink pads, scotch tape
Requires Fume hood, water bath, hair dryer, UV light (356nm, black light, strong sunlight, plant lights or a goggle cabinet), UV light short wave (254nm) and ring stands
Class Size 24 Students
Product Type Introduction to Toxicology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.