Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Hair Analysis
SCI_ED

Catalog No. S25728 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S25728 8 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard - Small parts not for children under three years.
Catalog No. S25728 Supplier Aqua Phoenix Scientific Supplier No. 10522
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard - Small parts not for children under three years.

Includes

Various hair samples, 35mm slides, cover slips, student diagram charts, all required chemicals, 24 student worksheets, teacher's manual, student instruction manuals

Requires

Microscopes

Study the morphology of hair samples from various species.

  • Uses slide sets to study the morphology of hair samples from various species
  • Isolate specific characteristics of the different types of hair
  • Prepare whole hair mounts as well as scale casts for making comparisons
  • Includes identification of unknowns
  • Grades 9 to 12
  • Class size: 24 students (six groups of four)

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) For study the morphology of hair samples
Includes Various hair samples, 35mm slides, cover slips, student diagram charts, all required chemicals, 24 student worksheets, teacher's manual, student instruction manuals
Requires Microscope, hair brushes, forceps, and plastic sandwich bags
Class Size 24 Students
Product Type Analysis of Hair Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.