Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Master Forensics Kit
SCI_ED

Catalog No. s25730 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
28 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S25730 28 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25730 Supplier Aqua Phoenix Scientific Supplier No. 10526
Only null left
Add to Cart
Add to Cart

Includes

Instructor maual, Student manual, MSDS, CD

Requires

Microscopes, UV light, hot plate, fume hood

Combine physics, chemistry and biology knowledge using forensics.

Introduces students to fingerprint, soil, hair, document and drugs and poison analysis.

  • Designed to build deductive powers and to focus on scientific method
  • Grades 7-14
  • Class size: 24 students (six groups of four)
Includes five units:
  • Analysis of Fingerprints Kit: Techniques for dusting and developing latent fingerprints from various surfaces
  • Analysis of Minerals and Soils Kit: Provides a number of residue samples for analysis
  • Hair Analysis Kit: Characterize specific aspects of hair from different species and in preparing specimens
  • Analysis of Drugs and Poisons Kit: Detect the presence of certain substances and discern the chemical and physical properties of writing materials
  • Analysis of Documents Kit: Use physical and chemical analysis to validate or repudiate a document

Specifications

Certifications/Compliance National standards
Grade 7 to 14
For Use With (Application) For designed to build deductive powers and to focus on the scientific method
Includes Five units are analysis of fingerprints kit, minerals and soils kit, hair analysis kit, drugs and poisons kit, documents kit
Class Size 28 Students
Product Type Master Forensics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.