Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ 96-Well Microplate
SCI_ED

Catalog No. S193841 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
12/Pk.
6/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S193841 12/Pk.
S19384 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S193841 Supplier Kemtec™ Supplier No. 4420B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for microchemistry experiments.

  • Clear acrylic
  • Measure 3.15 × 5.12 in. (8 × 13cm)
  • Durable and easy to clean

Specifications

Format Standard
Array 8 x 12
Color Clear
Material ABS Plastic
No. of Wells 96
For Use With (Application) Microchemistry experiments
Quantity 12/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.