Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Ocean Trash Can Kit
SCI_ED

Catalog No. s94963 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
7 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S94963 7 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S94963 Supplier Aqua Phoenix Scientific Supplier No. 9120
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor Manual
  • Student Data Sheets
  • Plastic Cups
  • Cupcake Liners
  • Sand
  • Tums
  • pH Strips
  • Radish Seeds
  • Sea Salt
  • Styrofoam Peanuts
  • Feathers
  • Yarn
  • Plastic Bags
  • Cotton Balls
  • Animal Fur
  • Wire

Requires

Six 1 gallon milk containers

Study effects of pollution on ocean, its inhabitants and surrounding coastline

This exercise examines various types of pollutants, their sources, and their effects on the ocean, its inhabitants and the surrounding coastline.

  • Students classify ocean pollution into five basic categories: debris, toxins, dissolved nutrients, particulates and oil
  • Activities include: effect of pH of liquids on calcium carbonate, effects of dilution, buffering capacity, salt and acid runoff, acid rain; ability of water to purify itself, and oil spill clean-up
  • Students design and write newsletter containing articles with pertinent pollution topics such as current laws, pollution, effects, bioaccumulation and prevention
  • Requires four class periods plus research time
  • Grades 7-10
  • Class size: 24 students

Specifications

Grade 7 to 10
For Use With (Application) Study effects of pollution on ocean, its inhabitants and surrounding coastline
Includes Instructor Manual; Student Data Sheets; Plastic Cups; Cupcake Liners; Sand; Tums; pH Strips; Radish Seeds; Sea Salt; Styrofoam Peanuts; Feathers; Yarn; Plastic Bags; Cotton Balls; Animal Fur; Wire
Requires Four class periods plus research time
Class Size 24 Students
Description Ocean Trash Can Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.