Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Osmosis and Diffusion Kit
SCI_ED

Catalog No. s06766 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06766 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06766 Supplier Aqua Phoenix Scientific Supplier No. 2302
Only null left
Add to Cart
Add to Cart

Includes

Instructor manual, student data sheets, methods of assessment manual, material safety data sheets, glucose test strips, dialysis tubing, pipets, two dram vials, iodine solution, glucose, starch

Requires

Standard laboratory supplies

Introduction to the relationship between solute and solvent, an interaction that is important to both plant and animal life.

Students will utilize starch and glucose solutions with dialysis tubing to model the cell membrane and observe the processes of osmosis and diffusion (essential concepts in understanding how cells survive).

  • Students can experiment with membranes and molecules
  • Illustrates cellular absorption in animal tissue
  • Helps to visualize exchange between water and solutes
  • Kit contains materials to carry out two complete experiments with glucose and starch
  • Three part lab takes no more than two class periods to complete
  • This laboratory activity is ideal for high school biology classes of all levels
  • For grades 9 to 12
  • Class size: 24 students

Specifications

Grade 8 to 12
Includes Instructor manual, student data sheets, methods of assessment manual, material safety data sheets, glucose test strips, dialysis tubing, pipets, two dram vials, iodine solution, glucose, starch
Product Type Osmosis and Diffusion Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.