Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Owl Pellet Exercise
SCI_ED

Catalog No. S02095 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Owl Pellet Kit
Owl Pellet Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S02095 Owl Pellet Refill
S02094 Owl Pellet Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S02095 Supplier Aqua Phoenix Scientific Supplier No. 1109R
Add to Cart
Add to Cart
This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

12 Owl pellets

Students discover what an owl had to eat by examining the owl pellet.

  • Pellet Refill: Includes 12 owl pellets
  • Students explore feeding habits of owls and their place in food chain
  • May be used by older students to study intricate balance and relationships between tropic levels in food web, energy flow and population densities
  • Grades: K to 12
  • Class size: 24 students

Specifications

For Use With (Application) Students discover what an owl had to eat by examining the owl pellets
Includes 12 Sterilized owl pellets
Product Type Owl Pellet Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.