Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Physical and Chemical Resources of the Living Ocean
SCI_ED

Catalog No. S94964 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S94964 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S94964 Supplier Aqua Phoenix Scientific Supplier No. 9124
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Requires

Refrigerator or freezer, hot plate, balance (x .01g), chloride/salinity test kit and laboratory glassware

Helps to learn why salt is one of our most valuable chemical resources.

This kit is designed to examine salt, not only as the most obvious property of seawater but also as one of its most valuable chemical resources.

  • Learn about varying salt concentrations of seawater and its amazingly uniform composition
  • Learn how salt can be removed from seawater
  • Experiments include: determining salinity by evaporation, determining salinity using a hydrometer, desalination by freezing, and desalination using a solar still
  • Allow five class periods
  • Target Grades: 9-12
  • Class size: 24 students

Specifications

Grade 9 to 12
For Use With (Application) Explore why salt is one of our most valuable chemical resources
Requires Five class periods
Class Size 24 Students
Description Physical and Chemical Resources of the Living Ocean Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.