Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Physical Properties of Proteins Kit
SCI_ED

Catalog No. S25811 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S25811 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25811 Supplier Aqua Phoenix Scientific Supplier No. 8152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructor Manual
  • Student Manual
  • Student Data Sheet
  • MSDS
  • Pipets
  • Empty Plastic Bottles
  • Ammonium Sulfate
  • Acetone Solution
  • Glacial Acetic Acid Solution
  • Lead Acetate Solution
  • Sodium Hydroxide
  • Albumin
  • Caseine
  • Gelatin

Requires

Goggles, Bunsen burner, glassware, test tube holders

Students learn how physical parameters influence proteins.

This kit's five experiments demonstrate the important effects physical parameters such as temperature, pH, salt concentration and dielectric constant have on proteins. Experiments range from the isoelectric precipitation of casein to effects of acetone on solutions of albumin and gelatin.

Kit can be used for following experiments:

  • Isoelectric precipitation of casein
  • Separation of albumin and gelatin by ammonium sulfate fractionation
  • Effects of acetone on albumin and gelatin solution
  • Heat denaturation of albumin
  • Precipitation of protein by lead acetate
  • Grades 9 to 14
  • Provides enough materials for 24 students working in groups of three

Contains lead.

Specifications

Grade 8 to 14
For Use With (Application) Demonstrates the important effects of physical parameters such as temperature, ph, salt concentration, and dielectric constants on proteins and illustrates how these effects are used in the isolation and purification of proteins
Includes Instructor manual, student manual, student data sheet, msds, pipettes, empty plastic bottles, ammonium sulfate, acetone solution, glacial acetic acid solution, lead acetate solution, sodium hydroxide, albumin, caseine, gelatin
Requires Bunsen burner, glassware, test tube holders
Class Size 24 Students
Product Type Physical Properties of Proteins Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.