Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Plant Cloning Kit
SCI_ED

Catalog No. s25020 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S25020 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S25020 Supplier Aqua Phoenix Scientific Supplier No. 2504
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Sterile Petri dishes
  • Bottles of silver nitrate
  • Seed-germinating media
  • Callus initiation media
  • Clone induction media
  • Carrot seeds
  • CD-ROM
  • Manual

Reproduce entire plant systems from a section of a single plant.

Propagating plants by cloning teaches the importance of sterile biological techniques.

  • Students practice sterile technique required for many biotechnology methods
  • Students germinate carrot seeds to provide plant material from which to establish callus culture
  • Students induce development of multiple shoots (plantlets) from callus tissue
  • Extended process takes approximately four months
  • No hood or incubator required
  • Includes detailed instruction manual and PowerPoint presentation with images of plant material at various stages of experiment
  • For grades 9 to 12
  • Class size: 24 students

Order Info

Perishable reply card items ship separately upon request and only in the continental US.

Specifications

Grade 8 to 12
For Use With (Application) Reproduce entire plant systems from a section of a single plant
Includes MSDS, methods of assessment manual, power point presentation with images of plant material at various stages of the experiment, Knife, spray bottle, pipets, shallow Petri dishes, deep Plastic bags, carrot seeds, plant cloning CD, seed germination media, callus initiation media, shoot induction media, Silver Nitrate solution, instructor manual
Time Range 4 months
Class Size 30 Students
Product Type Kemtec™ Plant Cloning-1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.