Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Qualitative Analysis of Proteins and Amino Acids Kit
SCI_ED

Catalog No. S25808 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S25808 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25808 Supplier Aqua Phoenix Scientific Supplier No. 8150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Instructor Manual
  • Student Manual
  • Student Data Sheets
  • MSDS
  • Pipet
  • Biuret Solution
  • Sodium Hypochlorite Solution
  • Alpha Naphthol Solution
  • Sodium Hydroxide (three different solutions)
  • Ninhydrin
  • Lead Acetate
  • Albumin
  • Gelatin
  • Glutamic Acid
  • Glycine
  • Cysteine
  • Arginine
  • DL Tryrosine
  • DL Tryptophane
  • Cystine
  • Unknow Amino Acid
  • Sodium Hydroxide

Requires

Goggles, concentrated nitric acid, ethanol, hot plates, balances, glassware and distilled water

Illustrates principles of protein and amino acid analysis.

Six experiments offer a wealth of scientific exploration and enable students to identify unknown compounds.

  • Students experience working with proteins (albumin and gelatin) and amino acids (glutamic acid, glycine, cystine, cysteine, tyrosine, tryptophan, and arginine)
  • Ninhydrin is used to detect presence of amines, indicating presence of amino acids and/or proteins
  • Biuret test is used to detect presence of peptide bonds
  • Specific analytical tests are used for determining presence of individual amino acids
  • For grades 10 to 14
  • Class size: 24 students

Specifications

Grade 10 to 14
For Use With (Application) Demonstrates the principles of protein and amino acid analysis and enables students to identify unknown compounds
Includes Instructor manual, student manual, student data sheets, MSDS, methods of assessment manual, Biuret Solution, Sodium Hypochlorite Solution, Alpha Naphthol solution, Sodium Hydroxide, Ninhydrin, Lead Acetate, Albumin, Gelatin, Glutamic Acid, Glycine, Cysteine, Arginine, DL Tryrosine, DL Tryptophane, Cystine, unknown Amino Acid, Sodium Hydroxide
Class Size 24 Students
Product Type Qualitative Analysis of Proteins and Amino Acids
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.