Learn More
Kemtec™ Tallquist Hemoglobin Scale
SCI_ED
Catalog No. S06452 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
8 to 14
Catalog No. Grade
S06452 8 to 14
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06452 Supplier Aqua Phoenix Scientific Supplier No. 12136
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.
Requires
Lancets if blood is to be drawn
Allows quick assessment of patient's blood hemoglobin levels.
Apply blood and allow to absorb into one of the test papers. Compare the color scale to determine the percent and weight of hemoglobin in blood under normal and anemic conditions.
  • Includes color chart and 150 test papers
  • Grade 9-14
Specifications
Grade 8 to 14
Includes Color chart and 150 test papers
Product Type Tallquist Hemoglobin Scale Kit