Learn More
Description
Includes
Instructor Manual, MSDS, Student Procedures, Student Data Sheet, Sucrose, Sodium Chloride, Quinine Sulfate, Vinegar, Empty Dropper Bottles
Requires
Labware for measuring taste test substances, which may be consumed
This kit features two lab experiments which can be completed in one hour or separated into two thirty minute activities
- Discover four taste sensations: sweet, sour, salt and bitter in blind taste test
- Students examine PTC inheritance, as well as their tendency to eat certain types of foods
- For learning about genetic inheritance or for studying human anatomy
- Grades 9-12
- Class size: 24 students
Specifications
Specifications
| Grade | 8 to 12 |
| For Use With (Application) | Provide students with physiological and genetic information about their bodies |
| Includes | Solutions of nacl, sucrose, quinine and acetic acid, 24 data sheets and manual |
| Product Type | Taste Discrimination Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
