Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Argyle™ Single Suction Catheters with Directional Valve

Catalog No. 22037930
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-037-930 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-037-930 Supplier Covidien Supplier No. 141902

Specifications

Material PVC
Color Green
For Use With (Application) Minimize the chance of exposure to aspirated sputum, maintains efficient suction with ease of insertion and withdrawal
Includes Directional control valve, 1cm graduation markings
Product Type Argyle™ Suction Catheter
Quantity 100/Cs.
Needle Point Style DeLee
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.