Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Kenpaullone
GREENER_CHOICESDP

Catalog No. 139810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-981-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-981-0 Supplier Tocris Bioscience™ Supplier No. 1398/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GSK-3β inhibitor; also inhibits cdks

GSK-3beta inhibitor (IC50 = 0.23 μM). Also inhibits cyclin dependent kinases (cdks) in the sub micromolar range (reported IC50 values are 0.4, 0.68-7 and 0.85 μM for cdk1, cdk2 and cdk5, respectively). Generates induced pluripotent stem cells (iPSCs) from somatic cells when used in combination with reprogramming factors; can replace Klf4.

Chemical Identifiers

CAS 142273-20-9
Molecular Formula C16H11BrN2O

Specifications

Quantity 10 mg
Formula Weight 327.18
Purity >98%
Molecular Weight (g/mol) 327.181
Chemical Name or Material Kenpaullone
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.