Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin, Powder, Spectrum™ Chemical
SDP

Catalog No. 18603631 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-631 5 g Poly Bottle
18-603-632 25 g Poly Bottle
18-603-633 100 g Poly Bottle
18-603-636 500 g Poly Bottle
18-603-635 2.5 kg Poly Bottle
18-603-634 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-631 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. K30305GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K3030

Spectrum™ Chemical Keratin, Powder is the key material that makes up the human body’s out layer of skin. It is also the key element of hair and nails. It is considered to be similar to chitin. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 69430-36-0
Molecular Formula C2H2BrClO2
Molecular Weight (g/mol) 173.39
InChI Key GEHJBWKLJVFKPS-UHFFFAOYNA-N
IUPAC Name 2-bromo-2-chloroacetic acid
SMILES OC(=O)C(Cl)Br

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 5 g
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Keratin, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.