Learn More
Sigma Aldrich Fine Chemicals Biosciences Kern PNJ series precision1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z742875
Encompass
The new standard in the laboratory with robust tuning fork measuring system.Automatic internal adjustment guarantees high degree of accuracy and makes the balance independent of its location of use. Ideal for mobile applications which require verification such as ambulatory gold and jewellery purchasingHigh-quality tuning fork measuring system for steady weight values and continuous weighingCapacity display A bar lights up to show how much of the weighing range is still availablePrecise counting The automatic reference weight optimisation of reference weight gradually improves the averag piece weight valueCompact size practical for small spacesLarge glass draught shield with 3 sliding doors for easy access to the items being weighed for models with weighing plate size A˜ 140 mm weighing space WxDxH 172x171x160 mm
Catalog No. 50-290-8638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.