Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, Low Odor Deodorized, Reagents

Catalog No. 01334871
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-871 4 L
01-334-874 1 L
01-334-872 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-871 Supplier Reagents Holdings Llc Supplier No. CK1900004C
Add to Cart
Add to Cart

Kerosene, Low Odor Deodorized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64742-48-9
Molecular Formula C12H26-C15H32
Synonym Paraffin oil

Specifications

Color Colorless to Slight Yellow
Boiling Point 175°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.767
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene, Low Odor Deodorized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.