Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene (Odorless), Fisher Chemical™

Catalog No. K1020 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
K1020 20 L Steel Pail
K10-4 4 L Glass Bottle
K10-200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. K1020
This item is not returnable. View return policy

Odorless

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-47-8
MDL Number MFCD00135561

Specifications

Melting Point -17.8°C
Color Yellow
Density 4.5 (Air = 1.0)
Boiling Point 151.1°C
Flash Point 71°C
Odor Odorless
Assay Percent Range >99%
Packaging Steel Pail
Quantity 20 L
UN Number 1223
Vapor Density 4.5g/mol
Vapor Pressure 5mmHg at 38°C
Physical Form Liquid
Chemical Name or Material Petroleum distillates,hydrotreated light, Odorless
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Wear personal protective equipment. Ensure adequate ventilation. Keep away from open flames, hot surfaces and sources of ignition. Do not breathe vapors or spray mist. Do not ingest. Avoid contact with skin and eyes. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:2
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.