Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, Petroleum Distillate, Spectrum™ Chemical
SDP

Catalog No. 18611157 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-157 20 L Steel Can
18-611-159 500 mL Amber Glass Bottle
18-611-155 1 L Amber Glass Bottle
18-611-156 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-157 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. K102520LTMT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8008-20-6

Specifications

Boiling Range 151° to 301°C
CAS Max % 1
Packaging Steel Can
Quantity 20 L
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.