Learn More
Apexbio Technology LLC 5-Aminofluorescein 3326-34-9 1g
SDP
Supplier:  Apexbio Technology LLC A46681000
Encompass_Preferred
5-Aminofluorescein (CAS 3326-34-9) is a fluorescent labeling reagent possessing an amino functional group that enables covalent conjugation to biomolecules particularly proteins 5-Aminofluorescein facilitates the monitoring of protein localization structural interactions and dynamic behavior in biological systems Based on these properties 5-Aminofluorescein holds research potential in fluorescence imaging analytical methods such as fluorescent microscopy flow cytometry fluorescence spectroscopy and studies of protein tracking protein protein interactions complex formation and investigation of molecular mechanisms in biomedical and cellular biology contexts
Catalog No. 50-000-93784
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.