Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ketoconazole, 98%

Catalog No. AC455470250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455470250 25 g
AC455470010 1 g
AC455470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455470250 Supplier Thermo Scientific Chemicals Supplier No. 455470250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ketoconazole, CAS # 65277-42-1, is a synthetic, broad-spectrum antifungal agent with the ability to inhibit a variety of fungal infections. | CAS: 65277-42-1 | C26H28Cl2N4O4 | 531.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ketoconazole is an imidazole antifungal agent
  • It functions by inhibiting synthesis of ergosterol, which is the fungal equivalent of cholesterol

Applications

  • Ketoconazole shows fungicide activity against a variety of yeast infections
  • It is an inhibitor of CYP3A4, which is necessary for the conversion of lanosterol to ergosterol
  • It increases fungal cellular permeability
  • It also inhibits endogenous respiration by disrupting membrane phospholipids
  • Ketoconazole can also inhibit the synthesis of thromboxane and sterols such as aldosterone, cortisol, and testosterone
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 531.44
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Melting Point 148.0°C to 150.0°C
Loss on Drying 0.5% max. (80°C, 4 h) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in methanol,less soluble in ethanol,slightly soluble in dichloromethane or chloroform
Formula Weight 531.44
Percent Purity 98%
Chemical Name or Material Ketoconazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not get in eyes,on skin,or on clothing.
Do not eat,drink or smoke
EINECSNumber 265-667-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.