accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNO(R)O(S)NNOClCl

CAS RN 65277-42-1

IUPAC Name: 1-[4-(4-{[(2S,4R)-2-(2,4-dichlorophenyl)-2-[(1H-imidazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]ethan-1-one
Synonyms: nizoral ketoconazole (substance) cis-ketoconazole ketoconazole preparation ketoderm (2S,4R)-ketoconazole KCZ nizoral A-D ketoconazol [inn-spanish] nizoral shampoo
Molecular Weight (g/mol): 531.43
Molecular Formula: C26H28Cl2N4O4
InChi Key: XMAYWYJOQHXEEK-ZEQKJWHPSA-N
SMILES: CC(=O)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=CN=C3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (6)
  • (3)
  • (9)
Percent Purity 
  • (2)
  • (7)
  • (6)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
  • (1)
  • (13)
Physical Form 
  • (2)
  • (7)
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (13)
  • (6)
  • (2)
  • (3)
  • (7)
  • (2)
  • (7)
  • (1)
  • (1)
  • (7)
  • (7)
  • (6)
  • (3)
  • (9)
  • (2)
  • (7)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (13)
  • (2)
  • (7)
  • (2)
  • (5)
  • (3)
  • (3)