Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoconazole, 99.17%, MP Biomedicals™

Catalog No. ICN15915890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15915890 500 mg
ICN15915850 50 mg
ICN15915880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15915890 Supplier MP Biomedicals, Inc Supplier No. 0215915890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym cis-1-Acetyl-4-(4-((2-(2, 4-dichlorophenyl)-2-(1H-1M-idazol-1-ylmethyl)-1, 3-dioxolan-4-yl) methoxy) phenyl)-piperazine, Ketoisdin
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Type Ketoconazole
Physical Form Powder
Color White
Quantity 500 mg
Assay Percent Range >99%
Formula Weight 531.44
Percent Purity 98%
Grade Not Available
Chemical Name or Material Ketoconazole
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.