Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603643 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-643 500 g Amber Glass Bottle
18-603-637 5 g Amber Glass Bottle
18-603-638 25 g Amber Glass Bottle
18-603-639 100 g Amber Glass Bottle
18-603-640 1 kg Amber Glass Bottle
18-603-641 2.5 kg Amber Glass Bottle
18-603-642 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-643 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. K1149500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K1149, 65277-42-1, C26H28Cl2N4O4

Spectrum™ Chemical Ketoconazole, USP is a synthetic antifungal medication that is used primarily to treat fungal infections. While its use as an oral tablet has been discontinued in many countries due to concerns that it caused serious liver injury, it is still used in a variety of topical administrations such as creams, which are used to treat tinea, candidal paronychia, and pityriasis versicolor and in shampoos used primarily to treat seborrheic dermatitis (dandruff). The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 531.43
InChI Key XMAYWYJOQHXEEK-ZEQKJWHPSA-N
IUPAC Name 1-[4-(4-{[(2S,4R)-2-(2,4-dichlorophenyl)-2-[(1H-imidazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]ethan-1-one
SMILES CC(=O)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=CN=C3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
UN Number UN2811
Specific Rotation -1° to +1°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Ketoconazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.