Learn More
Description
Ketoconazole USP is a white or almost white powder. Particle size distribution: 90% ≤ 9 μm.
Specifications
Specifications
| Chemical Name or Material | Ketoconazole USP (Micronized) |
| Packaging | Jar 60ml, White, HDPE, Round, 33-400, TRIVENI-DB, CT |
| CAS | 65277-42-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 531.4372 |
| Quantity | 10 g |
| Percent Purity | 99 to 101 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
