Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Ketoconazole USP (Micronized)
SDP

Catalog No. 14186450 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-450 1 kg
14-185-126 10 g
14-185-386 500 g
14-185-447 5 g
14-185-450 100 g
14-185-396 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-186-450 Supplier Medisca Supplier No. 027909
Add to Cart
Add to Cart

Ketoconazole USP is a white or almost white powder. Particle size distribution: 90% ≤ 9 μm.

Ketoconazole USP is a white or almost white powder. Particle size distribution: 90% ≤ 9 μm.

Specifications

Chemical Name or Material Ketoconazole USP (Micronized)
Packaging Jar 3800ml, White, HDPE, Round, 110-400, TRIVENI-DB, CT
CAS 65277-42-1
Type Drug Standard
Molecular Weight (g/mol) 531.4372
Quantity 1 kg
Percent Purity 99 to 101 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.