Learn More
α-Ketoglutaric acid disodium salt hydrate, ≥95%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 305-72-6 |
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Linear Formula | C5H4O5 Na2· xH2 O |
| Quantity | 100 g |
| Formula Weight | 190.06 (anhydrous basis) |
| Percent Purity | ≥95% |
| Physical Form | Neat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.