Learn More
Eurofins Discoverx α-Ketoglutaric acid sodium salt, 10 mg. (92-1212)
Supplier: Eurofins Discoverx 921212
α-Ketoglutaric acid is a naturally occurring ketone derivative of glutaric acid and intermediate of the Krebs cycle. It serves as a nitrogen transporter, antioxidant, anabolic, wound healer, and may have anti-aging properties. Used to treat kidney disease, GI disorders, bacterial infections, liver problems, cataracts, and yeast infections. Commonly sold as a supplement to help combat ammonia toxicity. Agonist of OXGR1. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.