Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketohexokinase Antibody (1020654) [mFluor Violet 500 SE], Novus Biologicals™
SDP

Catalog No. NB658946
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB658946 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB658946 Supplier R&D Systems Supplier No. FAB81771MFV500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Ketohexokinase Monoclonal antibody specifically detects Ketohexokinase in Human samples. It is validated for Western Blot,Immunocytochemistry

Specifications

Antigen Ketohexokinase
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1020654
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase)
Host Species Mouse
Immunogen E. coli-derived human Ketohexokinase, Met1-Val298, Accession # P50053
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3795
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.