Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoprofen 98.0+%, TCI America™
SDP

Catalog No. K0038250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K0038250G 250 g
K003825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K0038250G Supplier TCI America Supplier No. K0038250G

Chemical Identifiers

CAS 22071-15-4
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00055790
InChI Key DKYWVDODHFEZIM-UHFFFAOYSA-N
Synonym ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
PubChem CID 3825
ChEBI CHEBI:6128
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
SMILES CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 95°C
Color White
Quantity 250 g
UN Number 2811
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketoprofen
RTECSNumber UE7570000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.