Learn More
U.S. Pharmacopeia Ketorolac Tromethamine, 74103-07-4, MFCD00887595, 200 mg
Supplier:  U.S. Pharmacopeia 1356665
Encompass
Empirical Formula (Hill Notation): C15H13NO3 · C4H11NO3, Molecular Weight: 376.40, Synonym: (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Ketorolac tris salt, Toradol.
Catalog No. NC1088865
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.