Learn More
Apexbio Technology LLC JIB-04 199596-05-9 20mg
SDP
Supplier:  Apexbio Technology LLC B157920
Encompass_Preferred
JIB-04 (CAS 199596-05-9) is a selective small-molecule inhibitor targeting Jumonji histone demethylases specifically suppressing enzymes including JARID1A JMJD2 family members (JMJD2A-E) and JMJD3 Structurally categorized as a pyridine hydrazone derivative JIB-04 inhibits demethylase activity independently of competitive binding with -ketoglutarate In vitro assays revealed IC50 values ranging from 230 nM to 1100 nM across various Jumonji enzymes Additionally JIB-04 demonstrates preferential inhibition of cancer cell proliferation versus normal cells notably in lung and prostate cancer cell lines and effectively reduces tumor growth in mouse xenograft models highlighting its relevance for cancer research
Catalog No. 50-000-62006
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.