Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety KeyGuard™ Cool Coveralls with Hood
SDP

Catalog No. 19813027
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-027 2X-Large
19-813-023 Small
19-813-024 Medium
19-813-025 Large
19-813-026 X-Large
19-813-028 3X-Large
19-813-029 4X-Large
19-813-030 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-813-027 Supplier Keystone Safety Supplier No. KEYTECCOOL2XLWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microporous coverall with SMS back provides comfort and breathability in warm work environments.

  • Microporous coverall with SMS Back
  • Great for applications such as pharmaceutical, painting, spraying, industrial manufacturing, research, and clinical facilities
  • Breathability in warmer work environments
  • Elastic Wrist, Elastic Ankles
  • 100% Latex Free

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type Snap Front
Sterility Non-sterile
Latex Free Yes
Model KEY-TEC-COOL
Sleeve Style Long Sleeve
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.