Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety KeyGuard Microporous Lab Coats
SDP

Catalog No. 19070102
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-070-102 Large
19-070-100 Small
19-070-101 Medium
19-070-103 X-Large
19-070-104 2X-Large
19-070-105 3X-Large
19-070-106 4X-Large
19-070-107 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-070-102 Supplier Keystone Safety Supplier No. LC0WEKGLRG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Latex free microporous lab coats are a cost savings alternative to Tyvek and provide contamination control in a variety of environments including cleanrooms.

  • Non-linting
  • Microporous
  • Latex free
  • Cost savings alternative to Tyvek brand
  • Individually inspected for stitching, sizing, material and workmanship
  • Ideal for cleanroom environments including electronic, medical device, and pharmaceutical industries where garment control is a concern

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Gender Unisex
Material Microporous Film
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
Model LCO-WE-KG
Seam Type Serged
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
No. per Pack 30/Pk.
Construction Keyguard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.