Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats
SDP

Catalog No. 19068432
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-068-432 Large
19-068-431 Medium
19-068-434 2X-Large
19-068-433 X-Large
19-068-530 Small
19-068-535 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-068-432 Supplier Keystone Safety Supplier No. LC0WENWHDLRGWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.

Protect workers against dry particulates, dirt and grime

  • Non-woven, nonlinting, spunbond heavyweight polypropylene construction
  • Serged seams offer high tensile strength
  • Latex-free
  • Disposable
  • Elastic wrists, snap front, single collar, no pockets
  • Available in six sizes
  • Flame resistant

Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Collar Type Single Ply
Seam Type Serged
For Use With (Application) Electronics, Medical Device, Pharmaceutical
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.