Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Nylon Beard Nets, 1/16 in. Holes
SDP

Catalog No. 19055238
Change view
Click to view available options
Color:
Blue
White
Size:
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-055-238 Blue X-Large
19-055-239 Blue Large
19-130-3955 White Large
19-055-240 White X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-055-238 Supplier Keystone Safety Supplier No. 112HPIXLBLUE
This item is not returnable. View return policy

Soft honeycomb nylon beard nets provide flame-resistant protection.

  • Beard net
  • Soft heavyweight nylon, 1/16 in. hole
  • Honeycomb style
  • Flame-resistant
  • Available in large, x-large sizes
  • Available in white or blue color

Specifications

Type General Purpose Beard Cover
Size X-Large
Material Nylon
Color Blue
Disposable Single-use
For Use With (Application) Arc Flash Industry, Electrical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.